Learn More
Apexbio Technology LLC Cetirizine DiHCl 83881-52-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000088214
Encompass_Preferred
Cetirizine dihydrochloride (CAS 83881-52-1) is a second-generation antihistamine structurally related to hydroxyzine It functions as a racemic selective inverse agonist of the histamine H1 receptor thereby attenuating histamine-mediated cellular responses Owing to its low penetration of the blood-brain barrier cetirizine exhibits reduced sedative effects compared to first-generation antihistamines This compound is widely utilized in biomedical research to investigate allergic pathways histaminergic signaling and the pharmacological modulation of conditions such as seasonal allergic rhinitis angioedema and urticaria
Catalog No. 50-000-88214 Supplier Apexbio Technology LLC Supplier No. B15475.1
Cetirizine dihydrochloride (CAS 83881-52-1) is a second-generation antihistamine structurally related to hydroxyzine It functions as a racemic selective inverse agonist of the histamine H1 receptor thereby attenuating histamine-mediated cellular responses Owing to its low penetration of the blood-brain barrier cetirizine exhibits reduced sedative effects compared to first-generation antihistamines This compound is widely utilized in biomedical research to investigate allergic pathways histaminergic signaling and the pharmacological modulation of conditions such as seasonal allergic rhinitis angioedema and urticaria