Learn More
TARGETMOL CHEMICALS INC Cetirizine dihydrochloride
SDP
Catalog No. 502880772
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Cetirizine dihydrochloride (UCB P071) is an effective second-generation histamine H1 antagonist that is potent in the therapy of allergic rhinitis, confirmed urticaria, and pollen-induced asthma. Unlike other conventional antihistamines, it does not cause side effects of drowsiness or anticholinergic. Purity 100%
Catalog No. 50-288-0772 Supplier TARGETMOL CHEMICALS INC Supplier No. T0089500MG
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Cetirizine dihydrochloride (UCB P071) is an effective second-generation histamine H1 antagonist that is potent in the therapy of allergic rhinitis, confirmed urticaria, and pollen-induced asthma. Unlike other conventional antihistamines, it does not cause side effects of drowsiness or anticholinergic. Purity 100%