Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cetirizine impurity C dihydrochloride | 2702511-37-1 | 100.0% | 461.81 g/mol | C21H27Cl3N2O3 | 5 MG
SDP

Catalog No. 5000262239
Encompass_Preferred

Cetirizine Impurity C dihydrochloride is a characterized analytical reference standard used for impurity profiling and method development related to the antihistamine cetirizine. Supplied as a high-purity dihydrochloride salt in small quantities, it is intended for laboratory quality control, stability studies, and analytical verification.

  • High purity (99.95%) suitable for analytical standards and quality control
  • Available in small quantities for trace-level analysis and method development
  • Includes supporting documentation such as COA, SDS, and spectral data
  • Molecular formula C21H27Cl3N2O3 and molecular weight 461.81 g/mol
  • Appropriate for impurity profiling and stability testing of drug formulations

Catalog No. 50-002-62239 Supplier Medchemexpress LLC Supplier No. HY131256A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cetirizine Impurity C dihydrochloride is a characterized analytical reference standard used for impurity profiling and method development related to the antihistamine cetirizine. Supplied as a high-purity dihydrochloride salt in small quantities, it is intended for laboratory quality control, stability studies, and analytical verification.

  • High purity (99.95%) suitable for analytical standards and quality control
  • Available in small quantities for trace-level analysis and method development
  • Includes supporting documentation such as COA, SDS, and spectral data
  • Molecular formula C21H27Cl3N2O3 and molecular weight 461.81 g/mol
  • Appropriate for impurity profiling and stability testing of drug formulations

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.