Learn More
ATCC Streptomyces violaceoruber
Supplier:  ATCC BAA471
Encompass
Streptomyces violaceoruber strain John Innes Centre M145 is a prototrophic derivative of Streptomyces coelicolor A3(2), 1985, John Innes Centre.
Catalog No. 50-238-1604