Learn More
Aviva Systems Biology Corp CETP Antibody - N-terminal region (ARP51759_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51759_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CETP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CETP. Alternate names: BPIFF, HDLCQ10ApplicationWB. Species reactivity: Guinea Pig, Human, Rabbit, Rat
Catalog No. 50-014-60605