Learn More
Aviva Systems Biology Corp CETP Antibody - middle region (ARP51760_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51760_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CETP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CETP. Alternate names: BPIFF, HDLCQ10ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60606