Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cetraxate Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. C345450MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C345450MG 50 mg
C3454250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C345450MG Supplier TCI America Supplier No. C345450MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 27724-96-5
Molecular Formula C17H24ClNO4
Molecular Weight (g/mol) 341.83
MDL Number MFCD01722166
InChI Key USROQQUKEBHOFF-UHFFFAOYSA-N
Synonym 3-[4-[[trans-4-(Aminomethyl)cyclohexyl]carbonyloxy]phenyl]propanoic Acid Hydrochloride
PubChem CID 34001
IUPAC Name hydrogen 3-{4-[4-(aminomethyl)cyclohexanecarbonyloxy]phenyl}propanoic acid chloride
SMILES [H+].[Cl-].NCC1CCC(CC1)C(=O)OC1=CC=C(CCC(O)=O)C=C1

Specifications

Melting Point 240°C
Color White
Quantity 50 mg
Formula Weight 341.83
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cetraxate Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.