Learn More
MOLTOX Molecular Toxicology Inc Cetrimide Agar, 25 mL/plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948814
Encompass_Preferred
Also known as Pseudosel Agar, Cetrimide Agar is used as a selective isolation and identification media for Pseudomonas aeruginosa. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-814 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S014
Also known as Pseudosel Agar, Cetrimide Agar is used as a selective isolation and identification media for Pseudomonas aeruginosa. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.