Learn More
Alkali Scientific Cetrimide Agar, 20 mL Standard Monoplate Dish, 10/Pk
SDP
Catalog No. 5000662401
Encompass_Preferred
Cetrimide Agar, 20 mL Standard Monoplate Dish, 10/Pk is used for isolating Pseudomonas species in clinical and laboratory settings. The selective formulation supports the growth of Pseudomonas while inhibiting the growth of other microorganisms. Each 20mL monoplate is pre-prepared and sterile, ready to use directly from the package, making it an efficient choice for routine microbial analysis.
Catalog No. 50-006-62401 Supplier Alkali Scientific Supplier No. CETAP00
Cetrimide Agar, 20 mL Standard Monoplate Dish, 10/Pk is used for isolating Pseudomonas species in clinical and laboratory settings. The selective formulation supports the growth of Pseudomonas while inhibiting the growth of other microorganisms. Each 20mL monoplate is pre-prepared and sterile, ready to use directly from the package, making it an efficient choice for routine microbial analysis.