Learn More
Selleck Chemical LLC Cetrimonium Bromide (CTAB) S4242-1g
SDP
Catalog No. 503109973
Encompass_Preferred
Cetrimonium Bromide (CTAB) is a known component of the broad-spectrum antiseptic cetrimide which is a mixture of different quaternary ammonium salts
Catalog No. 50-310-9973 Supplier Selleck Chemical LLC Supplier No. S42421G
Cetrimonium Bromide (CTAB) is a known component of the broad-spectrum antiseptic cetrimide which is a mixture of different quaternary ammonium salts