Learn More
Medchemexpress LLC Cetrimonium bromide | 57-09-0 | 98.0% | 1 ML
SDP
Catalog No. 5000413251
Encompass_Preferred
Cetrimonium bromide (CTAB), a quaternary ammonium, is an orally active cationic surfactant. It exhibits toxicity and anticancer effects, inhibiting cell migration and invasion by modulating canonical and non-canonical TGF-β signaling pathways. Cetrimonium bromide is also utilized for DNA extraction.
  • Cationic surfactant.
  • Exhibits toxicity and anticancer effects.
  • Inhibits cell migration and invasion by modulating TGF-β signaling pathways.
  • Can be used for DNA extraction.
  • Inhibits the protein expression of MMP-2 and MMP-9, and increases TIMP-1 and TIMP-2 protein expression in SK-HEP-1 cells.
  • Induces apoptosis in human Head and neck cancer (HNC) cells.
  • Ablated tumor-forming capacity of FaDu cells and delayed growth of established tumors in FaDu cells tumor-bearing mice.
Catalog No. 50-004-13251 Supplier Medchemexpress LLC Supplier No. HYB126010MM/1ML
Add to Cart
Add to Cart
Cetrimonium bromide (CTAB), a quaternary ammonium, is an orally active cationic surfactant. It exhibits toxicity and anticancer effects, inhibiting cell migration and invasion by modulating canonical and non-canonical TGF-β signaling pathways. Cetrimonium bromide is also utilized for DNA extraction.
  • Cationic surfactant.
  • Exhibits toxicity and anticancer effects.
  • Inhibits cell migration and invasion by modulating TGF-β signaling pathways.
  • Can be used for DNA extraction.
  • Inhibits the protein expression of MMP-2 and MMP-9, and increases TIMP-1 and TIMP-2 protein expression in SK-HEP-1 cells.
  • Induces apoptosis in human Head and neck cancer (HNC) cells.
  • Ablated tumor-forming capacity of FaDu cells and delayed growth of established tumors in FaDu cells tumor-bearing mice.