Learn More
Medchemexpress LLC Cetrimonium bromide | 57-09-0 | 98.0% | 500 MG
SDP
Catalog No. 5000412835
Encompass_Preferred
Cetrimonium bromide (CTAB) is a quaternary ammonium, orally active cationic surfactant. It exhibits toxicity and anticancer effects. It inhibits cell migration and invasion by modulating canonical and non-canonical TGF-β signaling pathways. Cetrimonium bromide can also be used for DNA extraction.
  • Cationic surfactant
  • Exhibits toxicity and anticancer effects
  • Inhibits cell migration and invasion by modulating canonical and non-canonical TGF-β signaling pathways
  • Can be used for DNA extraction
  • Does not affect growth of SK-HEP-1 cells at 1-5 μM for 24 hours
  • Attenuates cellular migration and invasion of SK-HEP-1 cells at 5 μM for 16-24 hours
  • Inhibits protein expression of MMP-2 and MMP-9, increases TIMP-1 and TIMP-2, and restrains Rac1, cdc42, and RhoA in SK-HEP-1 cells at 5 μM for 24 hours
  • Induces apoptosis in human head and neck cancer (HNC) cells at 5 μM for 12-72 hours
Catalog No. 50-004-12835 Supplier Medchemexpress LLC Supplier No. HYB1260500MG
Cetrimonium bromide (CTAB) is a quaternary ammonium, orally active cationic surfactant. It exhibits toxicity and anticancer effects. It inhibits cell migration and invasion by modulating canonical and non-canonical TGF-β signaling pathways. Cetrimonium bromide can also be used for DNA extraction.
  • Cationic surfactant
  • Exhibits toxicity and anticancer effects
  • Inhibits cell migration and invasion by modulating canonical and non-canonical TGF-β signaling pathways
  • Can be used for DNA extraction
  • Does not affect growth of SK-HEP-1 cells at 1-5 μM for 24 hours
  • Attenuates cellular migration and invasion of SK-HEP-1 cells at 5 μM for 16-24 hours
  • Inhibits protein expression of MMP-2 and MMP-9, increases TIMP-1 and TIMP-2, and restrains Rac1, cdc42, and RhoA in SK-HEP-1 cells at 5 μM for 24 hours
  • Induces apoptosis in human head and neck cancer (HNC) cells at 5 μM for 12-72 hours