Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cetrimonium bromide-d42 | 907216-28-8 | 99.8% | 406.71 | C19D42BrN | 10 MG
SDP

Catalog No. 5000296756
Encompass_Preferred

Deuterium-labeled cetrimonium bromide (d42) is a stable isotope-labeled form of the cationic surfactant cetrimonium bromide, supplied for research use as a tracer or internal standard. It is provided as a powder at high purity with defined storage conditions.

  • Deuterium labeled (d42) tracer/internal standard.
  • Purity 99.8%.
  • Molecular formula C19D42BrN; molecular weight 406.71.
  • CAS number 907216-28-8.
  • Supplied as a powder in small pack sizes (5 mg, 10 mg, 25 mg; larger sizes by inquiry).
  • Storage: powder -20°C (3 years); in solvent -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-96756 Supplier Medchemexpress LLC Supplier No. HYB1260S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deuterium-labeled cetrimonium bromide (d42) is a stable isotope-labeled form of the cationic surfactant cetrimonium bromide, supplied for research use as a tracer or internal standard. It is provided as a powder at high purity with defined storage conditions.

  • Deuterium labeled (d42) tracer/internal standard.
  • Purity 99.8%.
  • Molecular formula C19D42BrN; molecular weight 406.71.
  • CAS number 907216-28-8.
  • Supplied as a powder in small pack sizes (5 mg, 10 mg, 25 mg; larger sizes by inquiry).
  • Storage: powder -20°C (3 years); in solvent -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.