Learn More
Medchemexpress LLC Cetrimonium-d9 bromide | 95217-14-4 | 50 MG
SDP
Catalog No. 5000412816
Encompass_Preferred
Cetrimonium-d9 (bromide) is the deuterium-labeled Cetrimonium bromide. Cetrimonium bromide (CTAB) is an amine-based cationic quaternary surfactant, and a component of the topical antiseptic Cetrimide. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12816 Supplier Medchemexpress LLC Supplier No. HYB1260S350MG
Cetrimonium-d9 (bromide) is the deuterium-labeled Cetrimonium bromide. Cetrimonium bromide (CTAB) is an amine-based cationic quaternary surfactant, and a component of the topical antiseptic Cetrimide. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.