Learn More
TARGETMOL CHEMICALS INC CETRORELIX ACETATE 10MG
SDP
Catalog No. 502579751
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cetrorelix Acetate is gonadotropin-releasing hormone (GnRH) receptor antagonist with an IC50 of 1.21 nM. purity: 97%
Catalog No. 50-257-9751 Supplier TARGETMOL CHEMICALS INC Supplier No. T552010MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cetrorelix Acetate is gonadotropin-releasing hormone (GnRH) receptor antagonist with an IC50 of 1.21 nM. purity: 97%