Learn More
Medchemexpress LLC Cetrorelix Acetate | 145672-81-7 | 98.25% | 1491.09 | 100 MG
SDP
Catalog No. 5000426352
Encompass_Preferred
Cetrorelix Acetate is a potent gonadotropin-releasing hormone (GnRH) receptor antagonist with an IC50 of 1.21 nM. It is also known as SB-75 acetate. This compound is used in research related to neurological diseases and endocrinology, and has shown antiproliferative effects on cell lines.
  • Potent GnRH receptor antagonist
  • Inhibits growth of ES-2 cell line
  • Applicable in neurological disease research
  • Applicable in endocrinology research
Catalog No. 50-004-26352 Supplier Medchemexpress LLC Supplier No. HYP0009A100MG
Cetrorelix Acetate is a potent gonadotropin-releasing hormone (GnRH) receptor antagonist with an IC50 of 1.21 nM. It is also known as SB-75 acetate. This compound is used in research related to neurological diseases and endocrinology, and has shown antiproliferative effects on cell lines.
  • Potent GnRH receptor antagonist
  • Inhibits growth of ES-2 cell line
  • Applicable in neurological disease research
  • Applicable in endocrinology research