Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cetrorelix acetate | 145672-81-7 | 98.3% | 1491.09 | 25 MG
SDP

Catalog No. 5000426764
Encompass_Preferred

Cetrorelix Acetate is a potent gonadotropin-releasing hormone (GnRH) receptor antagonist. It exhibits an IC50 of 1.21 nM and is utilized in various research applications, including studies on cancer cells and clinical trials for conditions such as polycystic ovarian syndrome and infertility.

  • Potent GnRH receptor antagonist
  • IC50 of 1.21 nM against GnRH
  • Inhibits growth of ES-2 cell line at 1000 ng/ml
  • Soluble in DMSO (50 mg/mL)
  • Soluble in H2O (2 mg/mL)
  • White to off-white solid appearance
  • Molecular formula C72H96ClN17O16
  • Long-term storage as powder at -80°C for 2 years or -20°C for 1 year
  • Long-term storage in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-26764 Supplier Medchemexpress LLC Supplier No. 25MGHYP0009A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cetrorelix Acetate is a potent gonadotropin-releasing hormone (GnRH) receptor antagonist. It exhibits an IC50 of 1.21 nM and is utilized in various research applications, including studies on cancer cells and clinical trials for conditions such as polycystic ovarian syndrome and infertility.

  • Potent GnRH receptor antagonist
  • IC50 of 1.21 nM against GnRH
  • Inhibits growth of ES-2 cell line at 1000 ng/ml
  • Soluble in DMSO (50 mg/mL)
  • Soluble in H2O (2 mg/mL)
  • White to off-white solid appearance
  • Molecular formula C72H96ClN17O16
  • Long-term storage as powder at -80°C for 2 years or -20°C for 1 year
  • Long-term storage in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.