Learn More
Medchemexpress LLC Cetrorelix Acetate | 145672-81-7 | 99.9% | 1491.09 | 50 MG
SDP
Catalog No. 5000426349
Encompass_Preferred
Cetrorelix Acetate is a potent gonadotropin-releasing hormone (GnRH) receptor antagonist with an IC50 of 1.21 nM. It is for research use only and inhibits the growth of the ES-2 cell line.
  • Potent GnRH receptor antagonist with an IC50 of 1.21 nM
  • Inhibits growth of ES-2 cell line at 1000 ng/ml
  • For research use only
Catalog No. 50-004-26349 Supplier Medchemexpress LLC Supplier No. HYP0009A50MG
Cetrorelix Acetate is a potent gonadotropin-releasing hormone (GnRH) receptor antagonist with an IC50 of 1.21 nM. It is for research use only and inhibits the growth of the ES-2 cell line.
  • Potent GnRH receptor antagonist with an IC50 of 1.21 nM
  • Inhibits growth of ES-2 cell line at 1000 ng/ml
  • For research use only