Learn More
Medchemexpress LLC Cetrorelix Acetate | 145672-81-7 | 99.92% | 1491.09 | 10 MG
SDP
Catalog No. 5000426518
Encompass_Preferred
Cetrorelix Acetate, also known as SB-75 acetate, is a potent gonadotropin-releasing hormone (GnRH) receptor antagonist with an IC50 of 1.21 nM. It is intended for research use only.
  • Inhibits the growth of ES-2 cell line at 1000 ng/mL
  • Shows comparable antiproliferative effects to GnRH-I agonists
  • Utilized in research for conditions such as Polycystic Ovarian Syndrome and Infertility
Catalog No. 50-004-26518 Supplier Medchemexpress LLC Supplier No. HYP0009A10MG
Cetrorelix Acetate, also known as SB-75 acetate, is a potent gonadotropin-releasing hormone (GnRH) receptor antagonist with an IC50 of 1.21 nM. It is intended for research use only.
  • Inhibits the growth of ES-2 cell line at 1000 ng/mL
  • Shows comparable antiproliferative effects to GnRH-I agonists
  • Utilized in research for conditions such as Polycystic Ovarian Syndrome and Infertility