Learn More
Cayman Chemical CetrorelIxacetate 10mg
SDP
Catalog No. 502994347
Encompass_Preferred
A synthetic peptide GnRHR antagonist (Kd = 0.2 nM); inhibits [D-Trp6]GnRH-induced activation of hGnRHR and a downstream luciferase reporter gene in murine LTK- cells (IC50 = 1.2 nM); reduces tumor volume and serum luteinizing hormone levels by 35% and 50% of control, respectively, in a human epithelial ovarian cancer mouse xenograft model at 60 μg per day; increases PMF survival in ovaries in cyclophosphamide treated mice at 0.5 mg/kg
Catalog No. 50-299-4347 Supplier Cayman Chemical Supplier No. 2391010 MG
A synthetic peptide GnRHR antagonist (Kd = 0.2 nM); inhibits [D-Trp6]GnRH-induced activation of hGnRHR and a downstream luciferase reporter gene in murine LTK- cells (IC50 = 1.2 nM); reduces tumor volume and serum luteinizing hormone levels by 35% and 50% of control, respectively, in a human epithelial ovarian cancer mouse xenograft model at 60 μg per day; increases PMF survival in ovaries in cyclophosphamide treated mice at 0.5 mg/kg