Learn More
Medchemexpress LLC HY-P9905 1mg Medchemexpress, Cetuximab CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP99051MG
Encompass_Preferred
Medchemexpress, HY-P9905 1mg Cetuximab CAS: Cetuximab is a monoclonal antibody that inhibits epidermal growth factor receptor ( EGFR ), with a K d of 0.201 nM for soluble EGFR by SPR. Cetuximab has potent antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4394