Learn More
Medchemexpress LLC HY-P9905 50mg , Cetuximab CAS:205923-56-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP990550MG
Encompass_Preferred
HY-P9905 50mg Cetuximab CAS: 205923-56-4 Cetuximab (C225) is a human IgG1 monoclonal antibody that inhibits epidermal growth factor receptor (EGFR), with a Kd of 0.201 nM for EGFR by SPR. Cetuximab has potent antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1220