Learn More
Medchemexpress LLC HY-P9905 5mg Medchemexpress, Cetuximab CAS:205923-56-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP99055MG
Encompass_Preferred
Medchemexpress, HY-P9905 5mg Cetuximab CAS:205923-56-4 Cetuximab (C225) is a monoclonal antibody that inhibits epidermal growth factor receptor (EGFR), with a Kd of 0.201 nM for soluble EGFR by SPR. Cetuximab has potent antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9434