Learn More
Medchemexpress LLC Cetuximab PBS 5mg | 5mg
SDP
Catalog No. 5000321980
Encompass_Preferred
Cetuximab (PBS) is a human IgG1 monoclonal antibody that can inhibit EGFR The SPR method measured the Kd value of Cetuximab for EGFR to be 0 201 nM Cetuximab has a highly effective anti-tumor effect[1]
Catalog No. 50-003-21980 Supplier Medchemexpress LLC Supplier No. HYP9S00085MG
Cetuximab (PBS) is a human IgG1 monoclonal antibody that can inhibit EGFR The SPR method measured the Kd value of Cetuximab for EGFR to be 0 201 nM Cetuximab has a highly effective anti-tumor effect[1]