Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Cetyl Alcohol
SCI_ED

Catalog No. S25675 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Solid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25675 100 g Solid
S25675A 500 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25675
Add to Cart
Add to Cart

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 36653-82-4
Molecular Formula C16H34O
Molecular Weight (g/mol) 242.45
MDL Number MFCD00004760
InChI Key BXWNKGSJHAJOGX-UHFFFAOYSA-N
Synonym 1-hexadecanol, cetyl alcohol, hexadecanol, cetanol, palmityl alcohol, hexadecyl alcohol, n-cetyl alcohol, cetaffine, cetylol, cetal
PubChem CID 2682
ChEBI CHEBI:16125
IUPAC Name hexadecan-1-ol
SMILES CCCCCCCCCCCCCCCCO

Specifications

Color White
Density 0.81g/mL
Packaging Solid
Linear Formula CH3(CH2)14CH2OH
Quantity 100 g
Formula Weight 242.43
Chemical Name or Material Cetyl Alcohol
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.