Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cetyl Alcohol, Solstice

Catalog No. 60044529 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-529 500 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-044-529 Supplier Solstice Supplier No. 03884500G

Tested according to Ph.Eur.

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36653-82-4
Molecular Formula C16H34O
Molecular Weight (g/mol) 242.45
MDL Number MFCD00004760
InChI Key BXWNKGSJHAJOGX-UHFFFAOYSA-N
Synonym 1-hexadecanol, cetyl alcohol, hexadecanol, cetanol, palmityl alcohol, hexadecyl alcohol, n-cetyl alcohol, cetaffine, cetylol, cetal
PubChem CID 2682
ChEBI CHEBI:16125
IUPAC Name hexadecan-1-ol
SMILES CCCCCCCCCCCCCCCCO

Specifications

Melting Point 49°C to 50°C
Boiling Point 190°C
Flash Point 170°C (338°F)
Packaging Poly bottle
Linear Formula CH3(CH2)15OH
Quantity 500 g
UN Number NONH for all modes of transport
Beilstein 1748475
Formula Weight 242.44g/mol
Chemical Name or Material Cetyl Alcohol, Tested According to Ph.Eur.
EINECSNumber 253-149-0
RTECSNumber MM0225000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.