Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cetyl trimethylammonium hydrogen sulfate, LiChropur™ EMD-Millipore

Catalog No. M1183130025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1183130025 25 g Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1183130025 Supplier MilliporeSigma Supplier No. 1.18313.0025

missing translation for 'validMainframeMsg'

Ion pair reagents for analytical HPLC LiChropur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68214-07-3
Molecular Formula C19H43NO4S
Molecular Weight (g/mol) 381.616
InChI Key UCRJJNVFJGKYQT-UHFFFAOYSA-M
Synonym cetyltrimethylammonium hydrogensulfate, hexadecyltrimethylammonium hydrogensulphate, 1-hexadecanaminium, n,n,n-trimethyl-, sulfate 1:1, hexadecyltrimethylammonium hydrogen sulfate, cetyltrimethylammonium hydrogen sulfate, n,n,n-trimethylhexadecan-1-aminium hydrogensulfate, hexadecyltrimethylammonium hydrogen sulphate, ammonium, hexadecyltrimethyl-, hydrogen sulfate, hexadecyltrimethylammonium bisulfate, n,n,n-trimethylhexadecan-1-aminium hydrogen sulfate 1:1:1
PubChem CID 109763
IUPAC Name hexadecyl(trimethyl)azanium;hydrogen sulfate
SMILES CCCCCCCCCCCCCCCC[N+](C)(C)C.OS(=O)(=O)[O-]

Specifications

Packaging Glass
Quantity 25 g
Formula Weight 381.61
Chemical Name or Material Cetyl trimethylammonium hydrogen sulfate
Recommended Storage Store at 15° to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.