Learn More
Medchemexpress LLC HY-B1464 500mg Medchemexpress, Cetylpyridinium (chloride) CAS:123-03-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1464500MG
Encompass_Preferred
Medchemexpress, HY-B1464 500mg Cetylpyridinium (chloride) CAS:123-03-5 Cetylpyridinium chloride, a cationic quaternary ammonium compound, is an anti-bacterial agent with broad-spectrum activity. Cetylpyridinium chloride is an effective anti-HBV capsid assembly inhibitor with an IC50 of 2.5 μM. Cetylpyridinium chloride is used in pesticides and various types of mouthwashes, and other personal care products. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2706