Learn More
Medchemexpress LLC Cetylpyridinium chloride | 123-03-5 | 99.8% | 1 ML
SDP
Catalog No. 5000413400
Encompass_Preferred
Cetylpyridinium chloride, a cationic quaternary ammonium compound, is an anti-bacterial agent with broad-spectrum activity. It is an effective anti-HBV capsid assembly inhibitor with an IC50 of 2.5 μM. It is used in pesticides and various types of mouthwashes, and other personal care products.
  • Cationic quaternary ammonium compound
  • Anti-bacterial agent with broad-spectrum activity
  • Effective anti-HBV capsid assembly inhibitor (IC50 of 2.5 μM)
  • Used in pesticides and various types of mouthwashes, and other personal care products
Catalog No. 50-004-13400 Supplier Medchemexpress LLC Supplier No. HYB146410MM/1ML
Cetylpyridinium chloride, a cationic quaternary ammonium compound, is an anti-bacterial agent with broad-spectrum activity. It is an effective anti-HBV capsid assembly inhibitor with an IC50 of 2.5 μM. It is used in pesticides and various types of mouthwashes, and other personal care products.
  • Cationic quaternary ammonium compound
  • Anti-bacterial agent with broad-spectrum activity
  • Effective anti-HBV capsid assembly inhibitor (IC50 of 2.5 μM)
  • Used in pesticides and various types of mouthwashes, and other personal care products