Learn More
Medchemexpress LLC Cetylpyridinium chloride monohydrate | 6004-24-6 | 99.8% | 1 ML
SDP
Catalog No. 5000412543
Encompass_Preferred
Cetylpyridinium chloride monohydrate is a cationic quaternary ammonium compound that functions as an antiseptic, effectively killing bacteria and other microorganisms. It is commonly used in various oral hygiene products to prevent plaque and reduce gingivitis. This compound also exhibits biological activity against viruses and in cellular models, and has been studied in clinical trials for conditions such as SARS-CoV-2 infection and periodontal diseases.
  • Cationic quaternary ammonium compound
  • Functions as an antiseptic
  • Kills bacteria and other microorganisms
  • Prevents plaque and reduces gingivitis
  • Used in mouthwashes, toothpastes, and throat/nasal sprays
  • Disrupts influenza viral envelope
  • Exhibits dose-dependent cytotoxicity in MDCK cells
  • Enhances survival and reduces influenza-associated morbidity and mortality in mice
Catalog No. 50-004-12543 Supplier Medchemexpress LLC Supplier No. HYB128910MM/1ML
Cetylpyridinium chloride monohydrate is a cationic quaternary ammonium compound that functions as an antiseptic, effectively killing bacteria and other microorganisms. It is commonly used in various oral hygiene products to prevent plaque and reduce gingivitis. This compound also exhibits biological activity against viruses and in cellular models, and has been studied in clinical trials for conditions such as SARS-CoV-2 infection and periodontal diseases.
  • Cationic quaternary ammonium compound
  • Functions as an antiseptic
  • Kills bacteria and other microorganisms
  • Prevents plaque and reduces gingivitis
  • Used in mouthwashes, toothpastes, and throat/nasal sprays
  • Disrupts influenza viral envelope
  • Exhibits dose-dependent cytotoxicity in MDCK cells
  • Enhances survival and reduces influenza-associated morbidity and mortality in mice