Learn More
Medchemexpress LLC Cetylpyridinium chloride monohydrate | 6004-24-6 | 99.8% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB1289500MG
Encompass_Preferred
Cetylpyridinium chloride monohydrate is a cationic quaternary ammonium compound utilized in various oral and nasal hygiene products like mouthwashes, toothpastes, and throat/nasal sprays. It acts as an antiseptic, effectively eliminating bacteria and other microorganisms. It also helps prevent plaque formation and reduces gingivitis.
  • Acts as an antiseptic that kills bacteria and other microorganisms.
  • Effectively prevents plaque.
  • Reduces gingivitis.
Catalog No. 50-004-12000