Learn More
Cayman Chemical Cetylpyridinium (chloride), Purity: ≥98%, CAS: 123-03-5, 100 mg
SDP
Catalog No. 502987191
Encompass_Preferred
A quaternary ammonium-containing cationic surfactant with broad-spectrum antiseptic activities; active against S. mutans, S. sanguis, E. coli, Oxford Staphylococcus, and C. albicans in media (MICs = 1.25-62.5 µg/ml); active against Oxford Staphylococcus in pooled human saliva (MICs = 7.8-15.6 µg/ml); slows plaque formation for at least 21 days in a rat incisor plaque model when applied topically at 0.025-2%
Catalog No. 50-298-7191 Supplier Cayman Chemical Supplier No. 21983100 MG
A quaternary ammonium-containing cationic surfactant with broad-spectrum antiseptic activities; active against S. mutans, S. sanguis, E. coli, Oxford Staphylococcus, and C. albicans in media (MICs = 1.25-62.5 µg/ml); active against Oxford Staphylococcus in pooled human saliva (MICs = 7.8-15.6 µg/ml); slows plaque formation for at least 21 days in a rat incisor plaque model when applied topically at 0.025-2%