Learn More
Crescent Chemical Co Inc CETYLTRIMETHYLAMMONIUM-BR
SDP
Catalog No. 50247420
Encompass_Preferred
Cetyltrimethylammonium-bromide cryst. pure Cationic surfactant used instead of SDS in electrophoresis of highly charged and membrane protein subunits (1). Surfactant for modifying silica for HPLC (2). Suitable for cell permeabilization (3) and DNA isolation (4). Increases the e
Catalog No. 50-247-420 Supplier Crescent Chemical Co Inc Supplier No. SE16530.02
Cetyltrimethylammonium-bromide cryst. pure Cationic surfactant used instead of SDS in electrophoresis of highly charged and membrane protein subunits (1). Surfactant for modifying silica for HPLC (2). Suitable for cell permeabilization (3) and DNA isolation (4). Increases the e