Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cetyltrimethylammonium Chloride Solution

Catalog No. 292737500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-273-7500ML 500 mL
29-273-725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 29-273-7500ML Supplier Sigma Aldrich Supplier No. 292737500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cetyltrimethylammonium chloride (CTAC) is a cationic surfactant that can be used in the synthesis of mesoporous molecular sieves and micelle complexes. It can also be used as a dispersant to prevent aggregation of nanoparticles.

Specifications

CAS 112-02-7
Density 0.968 g/mL at 25°C
Molecular Formula C19H42ClN
Refractive Index n20/D 1.3778
Linear Formula CH3(CH2)15N(Cl)(CH3)3
MDL Number MFCD00011773
Quantity 500 mL
Synonym Hexadecyltrimethylammonium chloride solution
Molecular Weight (g/mol) 320 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.