Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cevidoplenib dimesylate | 2043659-93-2 | 98.96% | 665.74 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY109082A50MG

Encompass_Preferred

Cevidoplenib dimesylate is an orally available inhibitor of spleen tyrosine kinase (Syk), possessing potential anti-inflammatory and immunomodulating activities. It is a solid, yellow to brown in color, and is shipped at room temperature.

  • Orally available Syk inhibitor
  • Potential anti-inflammatory and immunomodulating activities
  • High purity of 98.96%
  • Soluble in DMSO (25 mg/mL)
  • Storage conditions: 4°C sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-45246


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.