Learn More
Medchemexpress LLC Cevidoplenib dimesylate | 2043659-93-2 | 99.0% | 665.74 | 1 ML
SDP
Catalog No. 5000343587
Encompass_Preferred
Cevidoplenib is an orally available inhibitor of spleen tyrosine kinase (Syk), with potential anti-inflammatory and immunomodulating activities.
  • Orally available inhibitor of spleen tyrosine kinase (Syk).
  • Possesses potential anti-inflammatory activities.
  • Exhibits potential immunomodulating activities.
Catalog No. 50-003-43587 Supplier Medchemexpress LLC Supplier No. HY109082A100MG
Cevidoplenib is an orally available inhibitor of spleen tyrosine kinase (Syk), with potential anti-inflammatory and immunomodulating activities.
  • Orally available inhibitor of spleen tyrosine kinase (Syk).
  • Possesses potential anti-inflammatory activities.
  • Exhibits potential immunomodulating activities.