Learn More
Selleck Chemical LLC Cevimeline HCl hemihydrate
SDP
Catalog No. 5000004244
Encompass_Preferred
Cevimeline HCl hemihydrate(AF 102B SNI 2011 SNK 508) is a parasympathomimetic agent that act as an agonist at the muscarinic acetylcholine receptors M1 and M3
Catalog No. 50-000-04244 Supplier Selleck Chemical LLC Supplier No. S64325MG
Cevimeline HCl hemihydrate(AF 102B SNI 2011 SNK 508) is a parasympathomimetic agent that act as an agonist at the muscarinic acetylcholine receptors M1 and M3