Learn More
Medchemexpress LLC Cevimeline hydrochloride hemihydrate | 153504-70-2 | >98% | C10H17NOS.HCl.1/2H2O | 1 ML
SDP
Catalog No. 5000434034
Encompass_Preferred
Cevimeline hydrochloride hemihydrate is a quinuclidine derivative of acetylcholine and a selective muscarinic M1 and M3 receptor agonist. This orally active compound stimulates secretion by the salivary glands, making it a candidate therapeutic drug for xerostomia, including in Sjogren's syndrome. It is also known to cross the blood-brain barrier.
  • Selective muscarinic M1 and M3 receptor agonist
  • Stimulates salivary gland secretion
  • Can be used as a sialogogue for xerostomia
  • Crosses the blood-brain barrier
  • Targets mAChR
  • Affects gastrointestinal, urinary, and reproductive systems
  • Increases intracellular Ca2+ concentration in parotid cells (In vitro)
  • Induces salivation and increases blood flow in the parotid gland (In vivo)
Catalog No. 50-004-34034 Supplier Medchemexpress LLC Supplier No. HY76772B10MM/1ML
Cevimeline hydrochloride hemihydrate is a quinuclidine derivative of acetylcholine and a selective muscarinic M1 and M3 receptor agonist. This orally active compound stimulates secretion by the salivary glands, making it a candidate therapeutic drug for xerostomia, including in Sjogren's syndrome. It is also known to cross the blood-brain barrier.
  • Selective muscarinic M1 and M3 receptor agonist
  • Stimulates salivary gland secretion
  • Can be used as a sialogogue for xerostomia
  • Crosses the blood-brain barrier
  • Targets mAChR
  • Affects gastrointestinal, urinary, and reproductive systems
  • Increases intracellular Ca2+ concentration in parotid cells (In vitro)
  • Induces salivation and increases blood flow in the parotid gland (In vivo)