Learn More
Medchemexpress LLC Cevimeline hydrochloride hemihydrate | 153504-70-2 | 99.5% | C10H17NOS.HCl.1/2H2O | 1 ML
SDP
Catalog No. 5000434063
Encompass_Preferred
Cevimeline hydrochloride hemihydrate is a quinuclidine derivative of acetylcholine and a selective, orally active muscarinic M1 and M3 receptor agonist. It stimulates secretion by the salivary glands, serving as a sialogogue for xerostomia, and can cross the blood-brain barrier.
  • Selective muscarinic M1 and M3 receptor agonist
  • Orally active
  • Stimulates salivary gland secretion
  • Used as a sialogogue for xerostomia
  • Crosses the blood-brain barrier
Catalog No. 50-004-34063 Supplier Medchemexpress LLC Supplier No. HY7677210MM/1ML
Cevimeline hydrochloride hemihydrate is a quinuclidine derivative of acetylcholine and a selective, orally active muscarinic M1 and M3 receptor agonist. It stimulates secretion by the salivary glands, serving as a sialogogue for xerostomia, and can cross the blood-brain barrier.
  • Selective muscarinic M1 and M3 receptor agonist
  • Orally active
  • Stimulates salivary gland secretion
  • Used as a sialogogue for xerostomia
  • Crosses the blood-brain barrier