Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cevimeline hydrochloride (synonyms: AF102B hydrochloride) | 107220-28-0 | 98.7% | C10H18ClNOS | 1 ML
SDP

Catalog No. 5000434720
Encompass_Preferred

Cevimeline hydrochloride is a quinuclidine derivative of acetylcholine and a selective and orally active muscarinic M1 and M3 receptor agonist. It stimulates secretion by the salivary glands and can be used as a sialogogue for xerostomia. It can also cross the blood-brain barrier.

  • Quinuclidine derivative of acetylcholine
  • Selective and orally active muscarinic M1 and M3 receptor agonist
  • Stimulates secretion by salivary glands
  • Used as a sialogogue for xerostomia
  • Can cross the blood-brain barrier
  • For research use only
  • Molecular weight: 235.77
  • Appearance: white to off-white solid
  • Soluble in H2O at concentrations ≥ 50 mg/mL

Catalog No. 50-004-34720 Supplier Medchemexpress LLC Supplier No. HY70020B10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cevimeline hydrochloride is a quinuclidine derivative of acetylcholine and a selective and orally active muscarinic M1 and M3 receptor agonist. It stimulates secretion by the salivary glands and can be used as a sialogogue for xerostomia. It can also cross the blood-brain barrier.

  • Quinuclidine derivative of acetylcholine
  • Selective and orally active muscarinic M1 and M3 receptor agonist
  • Stimulates secretion by salivary glands
  • Used as a sialogogue for xerostomia
  • Can cross the blood-brain barrier
  • For research use only
  • Molecular weight: 235.77
  • Appearance: white to off-white solid
  • Soluble in H2O at concentrations ≥ 50 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.