Learn More
Medchemexpress LLC Cevipabulin fumarate | 849550-67-0 | 99.9% | 580.89 | 25 MG
SDP
Catalog No. 5000361257
Encompass_Preferred
Cevipabulin fumarate is an oral, microtubule-active, antitumor compound that inhibits the binding of [3H]NSC 49842 to tubulin. It exhibits an IC50 of 18-40 nM for cytotoxicity in human tumor cell lines, demonstrating good activity on various cancer cell lines.
  • Oral, microtubule-active, antitumor compound
  • Inhibits binding of [3H]NSC 49842 to tubulin
  • Shows cytotoxicity in human tumor cell lines (IC50 of 18-40 nM)
  • Active against ovarian, breast, prostate, and cervical tumors
  • Produces sub-G1 nuclei at low concentrations (20-40 nM)
  • Causes strong G2-M block at concentrations above 50 nM
  • Active in vivo against human tumor xenografts
  • Exhibits dose-dependent antitumor activity
  • Targets microtubule/tubulin and cell cycle/DNA damage pathways
Catalog No. 50-003-61257 Supplier Medchemexpress LLC Supplier No. 25MGHY14949C
Cevipabulin fumarate is an oral, microtubule-active, antitumor compound that inhibits the binding of [3H]NSC 49842 to tubulin. It exhibits an IC50 of 18-40 nM for cytotoxicity in human tumor cell lines, demonstrating good activity on various cancer cell lines.
  • Oral, microtubule-active, antitumor compound
  • Inhibits binding of [3H]NSC 49842 to tubulin
  • Shows cytotoxicity in human tumor cell lines (IC50 of 18-40 nM)
  • Active against ovarian, breast, prostate, and cervical tumors
  • Produces sub-G1 nuclei at low concentrations (20-40 nM)
  • Causes strong G2-M block at concentrations above 50 nM
  • Active in vivo against human tumor xenografts
  • Exhibits dose-dependent antitumor activity
  • Targets microtubule/tubulin and cell cycle/DNA damage pathways