Learn More
Medchemexpress LLC Cevipabulin fumarate | 849550-67-0 | 99.9% | 580.89 | 50 MG
SDP
Catalog No. 5000360803
Encompass_Preferred
Cevipabulin fumarate (TTI-237 fumarate) is an oral, microtubule-active, antitumor compound. It inhibits the binding of [3H]NSC 49842 to tubulin, with an IC50 of 18-40 nM for cytotoxicity in human tumor cell lines.
  • Microtubule-active, antitumor compound
  • Shows good activity (18-40 nM IC50 values) on ovarian, breast, prostate, and cervical tumor cell lines
  • Produces sub-G1 nuclei at low concentrations (20-40 nM)
  • Causes a strong G2-M block at concentrations above 50 nM
  • Active via intravenous (i.v.) and oral (p.o.) administration against human tumor xenografts in mice, with good antitumor activity at 20 and 15 mg/kg
Catalog No. 50-003-60803 Supplier Medchemexpress LLC Supplier No. HY14949C50MG
Cevipabulin fumarate (TTI-237 fumarate) is an oral, microtubule-active, antitumor compound. It inhibits the binding of [3H]NSC 49842 to tubulin, with an IC50 of 18-40 nM for cytotoxicity in human tumor cell lines.
  • Microtubule-active, antitumor compound
  • Shows good activity (18-40 nM IC50 values) on ovarian, breast, prostate, and cervical tumor cell lines
  • Produces sub-G1 nuclei at low concentrations (20-40 nM)
  • Causes a strong G2-M block at concentrations above 50 nM
  • Active via intravenous (i.v.) and oral (p.o.) administration against human tumor xenografts in mice, with good antitumor activity at 20 and 15 mg/kg