Learn More
Medchemexpress LLC Cevipabulin fumarate | 849550-67-0 | MFCD31813783 | 99.9% | 580.89 g/mol | C22H22ClF5N6O5 | 10 MG
SDP
Catalog No. 5000286064
Encompass_Preferred
Cevipabulin fumarate is the fumarate salt of a microtubule-active small-molecule antitumor agent supplied for laboratory research use. It inhibits tubulin binding, disrupts microtubule dynamics, and produces cytotoxic effects in human tumor cell lines; supplied as a solid with specified storage conditions for stability.
  • Microtubule-active mechanism that disrupts tubulin dynamics.
  • High chemical purity suitable for research applications.
  • Solid, white to off-white physical form for easy handling and storage.
  • Storage conditions provided for solid and solution forms to preserve stability.
  • Available in small research pack sizes for preclinical studies.
Catalog No. 50-002-86064 Supplier Medchemexpress LLC Supplier No. HY14949C10MG
Cevipabulin fumarate is the fumarate salt of a microtubule-active small-molecule antitumor agent supplied for laboratory research use. It inhibits tubulin binding, disrupts microtubule dynamics, and produces cytotoxic effects in human tumor cell lines; supplied as a solid with specified storage conditions for stability.
  • Microtubule-active mechanism that disrupts tubulin dynamics.
  • High chemical purity suitable for research applications.
  • Solid, white to off-white physical form for easy handling and storage.
  • Storage conditions provided for solid and solution forms to preserve stability.
  • Available in small research pack sizes for preclinical studies.