Learn More
Medchemexpress LLC Cevipabulin fumarate (TTI-237 fumarate) | 849550-67-0 | 99.9% | 580.89 | 1 ML
SDP
Catalog No. 5000360269
Encompass_Preferred
Cevipabulin fumarate (TTI-237 fumarate) is an oral, microtubule-active, antitumor compound. It inhibits the binding of [3H]NSC 49842 to tubulin, with an IC50 of 18-40 nM for cytotoxicity in human tumor cell lines. It is for research use only.
  • Inhibits tubulin binding with IC50 values of 18-40 nM in human tumor cells.
  • Active against ovarian, breast, prostate, and cervical tumor cell lines.
  • Induces sub-G1 nuclei at 20-40 nM and G2-M block above 50 nM.
  • Demonstrates in vitro cytotoxicity (IC50 18-40 nM) in human cancer cell lines.
  • Active in vivo via i.v. and p.o. administration against human tumor xenografts.
  • Shows dose-dependent antitumor activity in mouse xenograft models.
Catalog No. 50-003-60269 Supplier Medchemexpress LLC Supplier No. HY14949C10MM/1ML
Cevipabulin fumarate (TTI-237 fumarate) is an oral, microtubule-active, antitumor compound. It inhibits the binding of [3H]NSC 49842 to tubulin, with an IC50 of 18-40 nM for cytotoxicity in human tumor cell lines. It is for research use only.
  • Inhibits tubulin binding with IC50 values of 18-40 nM in human tumor cells.
  • Active against ovarian, breast, prostate, and cervical tumor cell lines.
  • Induces sub-G1 nuclei at 20-40 nM and G2-M block above 50 nM.
  • Demonstrates in vitro cytotoxicity (IC50 18-40 nM) in human cancer cell lines.
  • Active in vivo via i.v. and p.o. administration against human tumor xenografts.
  • Shows dose-dependent antitumor activity in mouse xenograft models.