Learn More
TARGETMOL CHEMICALS INC CEVOGLITAZAR 5MG
Supplier: TARGETMOL CHEMICALS INC T269865MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Cevoglitazar (LBM-642) is a PPARɑ agonist and PPAR(gamma) agonist. Cevoglitazar potently reduces food intake and body weight in obese mice and cynomolgus monkeys. purity: 97%