Learn More
QA Bio Inc LudgerClean CEX cartridges - 6 cartridges
Catalog No. 502236697
Encompass
Cation exchange resin cartridges • Developed for the purification of O-glycans • Each pack contains 6 cartridges.
This item is not returnable. View return policy
Catalog No. 50-223-6697 Supplier QA Bio Inc Supplier No. LCCEXA6
Cation exchange resin cartridges • Developed for the purification of O-glycans • Each pack contains 6 cartridges.