Learn More
Stellar Solutions Inc CitriSurf 2310 Gel | 1 Gal
SDP
Catalog No. NC1731530
Encompass
CitriSurf 2310 is a high quality blend of chemicals formulated for cleaning, rust removal and passivation of stainless steel products. It is provided in a high viscosity form for easy application on weld joints, vertical surfaces, and other large items or areas that cannot be conveniently dipped in a bath. CitriSurf 2310 is specifically designed to provide low cost and efficient removal of all rust and free iron from the surface of stainless steel and other metals, providing the highest chrome oxide levels possible.
Catalog No. NC1731530 Supplier Stellar Solutions Inc Supplier No. CF12310
CitriSurf 2310 is a high quality blend of chemicals formulated for cleaning, rust removal and passivation of stainless steel products. It is provided in a high viscosity form for easy application on weld joints, vertical surfaces, and other large items or areas that cannot be conveniently dipped in a bath. CitriSurf 2310 is specifically designed to provide low cost and efficient removal of all rust and free iron from the surface of stainless steel and other metals, providing the highest chrome oxide levels possible.