Learn More
Norgen Biotek Corp cf-DNA/cf-RNA Preservative Tubes, 50 Pack
Catalog No. NC1715829
Encompass_Preferred
Norgen's cf-DNA/cf-RNA Preservative Tubes are closed, evacuated plastic tubes for the collection and the preservation of cf-DNA, circulating tumor DNA, cf-RNA and circulating tumor cells in human whole blood samples during storage and shipping. Using these tubes in combination with Norgen's Plasma/Serum Cell-Free Circulating DNA Purification Kits or Norgen's Plasma/Serum RNA Purification Kits enables the purification and concentration of an inhibitor-free cf-DNA/ct-DNA/cf-RNA in a very small elution volume. Plasma recovered from Norgen's cf-DNA/cf-RNA Preservative Tubes is also compatible with any other cf-DNA and/or cf-RNA purification method. The purified cf-DNA, ct-DNA and cf-RNA are compatible with any downstream application assays, including PCR, qPCR, Southern Blot analysis, gene expression analysis, microarrays and NGS.This product is for research use only and not for use in diagnostic procedures.
Catalog No. NC1715829 Supplier Norgen Biotek Corp Supplier No. 63950
Add to Cart
Add to Cart
Norgen's cf-DNA/cf-RNA Preservative Tubes are closed, evacuated plastic tubes for the collection and the preservation of cf-DNA, circulating tumor DNA, cf-RNA and circulating tumor cells in human whole blood samples during storage and shipping. Using these tubes in combination with Norgen's Plasma/Serum Cell-Free Circulating DNA Purification Kits or Norgen's Plasma/Serum RNA Purification Kits enables the purification and concentration of an inhibitor-free cf-DNA/ct-DNA/cf-RNA in a very small elution volume. Plasma recovered from Norgen's cf-DNA/cf-RNA Preservative Tubes is also compatible with any other cf-DNA and/or cf-RNA purification method. The purified cf-DNA, ct-DNA and cf-RNA are compatible with any downstream application assays, including PCR, qPCR, Southern Blot analysis, gene expression analysis, microarrays and NGS.This product is for research use only and not for use in diagnostic procedures.