Learn More
Maine Molecular Quality Controls Inc INTROL CF Panel III Control
SDP
Catalog No. NC1432223
Encompass
G115-1 - INTROL CF Panel III Control is intended for in vitro use as a quality control to monitor analytical performance of the extraction, amplification and detection steps of systems used in the qualitative measurement of the Cystic Fibrosis Transmembrane Conductance Regulator (CFTR) gene. This product is intended to be extracted and analyzed routinely with each cystic fibrosis (CF) test run. INTROL CF Panel III is designed to monitor the detection of 79 CFTR mutations, genetic risk factors associated with cystic fibrosis, including the 23 mutations recommended by ACMG and ACOG. INTROL CF Panel III Control also monitors variants I506V, I507V, F508C, 5/7/9/11T and I148T. INTROL CF Panel III is provided for Research Use Only (RUO). G115 contains: 4 bottles, 2 milliliters each.
Catalog No. NC1432223 Supplier Maine Molecular Quality Controls Inc Supplier No. G115
G115-1 - INTROL CF Panel III Control is intended for in vitro use as a quality control to monitor analytical performance of the extraction, amplification and detection steps of systems used in the qualitative measurement of the Cystic Fibrosis Transmembrane Conductance Regulator (CFTR) gene. This product is intended to be extracted and analyzed routinely with each cystic fibrosis (CF) test run. INTROL CF Panel III is designed to monitor the detection of 79 CFTR mutations, genetic risk factors associated with cystic fibrosis, including the 23 mutations recommended by ACMG and ACOG. INTROL CF Panel III Control also monitors variants I506V, I507V, F508C, 5/7/9/11T and I148T. INTROL CF Panel III is provided for Research Use Only (RUO). G115 contains: 4 bottles, 2 milliliters each.