Learn More
Sterlitech Corporation CF016A CELL ASSEMBLY CROSSFLOW
SDP
Supplier:  Sterlitech Corporation 1160310
Encompass_Preferred
CF016 Cell Assembly, Crossflow, Acrylic (CF016A). The Sterlitech CF016 membrane cell is a lab-scale cross flow filtration unit designed to provide fast and accurate performance data with minimal amounts of product, expense, and time.
Catalog No. NC2272187